ENA388700447 Enamine

1-(aminomethyl)-7-methoxy-1,2,3,4-tetrahydronaphthalen-1-ol hydrochloride

  • MDL number MFCD23144203
  • Cas Number : 1427381-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124400
Availability details for
Quantity
  • Availability
  • Add to Cart