ENA388700454 Enamine

(4,4-dimethyl-1,2,3,4-tetrahydronaphthalen-1-yl)methanamine

  • MDL number MFCD11641547
  • Cas Number : 1096815-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124421

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124421
Availability details for
Quantity
  • Availability
  • Add to Cart