ENA388700455 Enamine

1,2-dimethylcyclopropan-1-ol

  • MDL number MFCD21849666
  • Cas Number : 140401-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124422

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124422
Availability details for
Quantity
  • Availability
  • Add to Cart