ENA388700461 Enamine

2,2-difluoro-2-[2-(trifluoromethyl)phenyl]acetic acid

  • MDL number MFCD17223370
  • Cas Number : 1343107-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122741
Availability details for
Quantity
  • Availability
  • Add to Cart