ENA388700468 Enamine

6-phenyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2-carbaldehyde

  • MDL number MFCD17408051
  • Cas Number : 1272811-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124441
Availability details for
Quantity
  • Availability
  • Add to Cart