ENA388700471 Enamine

1-phenyl-N-[2-({1-phenyl-1H,4H,5H,6H-cyclopenta[c]pyrazol-3-yl}formamido)ethyl]-1H,4H,5H,6H-cyclopenta[c]pyrazole-3-carboxamide

  • MDL number MFCD23144206
  • Cas Number : 1427380-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124446
Availability details for
Quantity
  • Availability
  • Add to Cart