ENA388700472 Enamine

3,3-dimethyl-1-[2-(methylamino)ethyl]urea

  • MDL number MFCD21809909
  • Cas Number : 1423117-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124448
Availability details for
Quantity
  • Availability
  • Add to Cart