ENA388700474 Enamine

6-amino-1-[(4-methylcyclohexyl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD19123544
  • Cas Number : 1271479-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122750
Availability details for
Quantity
  • Availability
  • Add to Cart