ENA388700475 Enamine

2-(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)acetic acid

  • MDL number MFCD08700556
  • Cas Number : 1007515-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-122752

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122752
Availability details for
Quantity
  • Availability
  • Add to Cart