ENA388700480 Enamine

bicyclo[4.2.0]octa-1(6),2,4-trien-7-ol

  • MDL number MFCD00798161
  • Cas Number : 35447-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-60
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122758
Availability details for
Quantity
  • Availability
  • Add to Cart