ENA388700485 Enamine

2-amino-N-methylcyclopentane-1-carboxamide

  • MDL number MFCD20424880
  • Cas Number : 1341180-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122846
Availability details for
Quantity
  • Availability
  • Add to Cart