ENA388700490 Enamine

1-(2,6-dimethylphenyl)ethan-1-ol

  • MDL number MFCD00016856
  • Cas Number : 19447-06-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122863
Availability details for
Quantity
  • Availability
  • Add to Cart