ENA388700491 Enamine

2-amino-2-(3-chlorophenyl)acetic acid hydrochloride

  • MDL number MFCD06410535
  • Cas Number : 1214196-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-122864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122864
Availability details for
Quantity
  • Availability
  • Add to Cart