ENA388700495 Enamine

2-ethyl-4-methylcyclohexan-1-one

  • MDL number MFCD16091208
  • Cas Number : 115684-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124525
Availability details for
Quantity
  • Availability
  • Add to Cart