ENA388700497 Enamine

ethyl 2-(tert-butoxy)acetate

  • MDL number MFCD11196077
  • Cas Number : 42415-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124528
Availability details for
Quantity
  • Availability
  • Add to Cart