ENA388700500 Enamine

5-(4-methylphenoxymethyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD23144030
  • Cas Number : 1423626-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122869
Availability details for
Quantity
  • Availability
  • Add to Cart