ENA388700501 Enamine

ethyl 2-sulfamoylacetate

  • MDL number MFCD12783410
  • Cas Number : 55897-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-68
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124537
Availability details for
Quantity
  • Availability
  • Add to Cart