ENA388700504 Enamine

4-hydroxy-3-nitro-1,2-dihydroquinolin-2-one

  • MDL number MFCD00100916
  • Cas Number : 15151-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124540
Availability details for
Quantity
  • Availability
  • Add to Cart