ENA388700505 Enamine

3-methoxy-4-nitrobenzaldehyde

  • MDL number MFCD08703367
  • Cas Number : 80410-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124541

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124541
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart