ENA388700507 Enamine

1-(2-bromoethoxy)-2-chloro-4-fluorobenzene

  • MDL number MFCD12813775
  • Cas Number : 67160-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124543
Availability details for
Quantity
  • Availability
  • Add to Cart