ENA388700520 Enamine

3,3-dimethylcyclobutan-1-amine hydrochloride

  • MDL number MFCD22987614
  • Cas Number : 1284247-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124559
Availability details for
Quantity
  • Availability
  • Add to Cart