ENA388700526 Enamine

1-(2-methoxyethyl)cyclopentan-1-amine

  • MDL number MFCD16818538
  • Cas Number : 896734-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-122872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122872
Availability details for
Quantity
  • Availability
  • Add to Cart