ENA388700527 Enamine

2-sulfanylacetohydrazide

  • MDL number MFCD18263132
  • Cas Number : 760-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122874

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-122874
pictograms GHS06,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart