ENA388700531 Enamine

3-(2-fluorophenyl)-2,5-dihydro-1H-pyrrole hydrochloride

  • MDL number MFCD20687860
  • Cas Number : 1239205-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124570
Availability details for
Quantity
  • Availability
  • Add to Cart