ENA388700532 Enamine

3-(4-hydroxyphenyl)-3-oxopropanoic acid

  • MDL number MFCD20653742
  • Cas Number : 62024-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124573

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124573
Availability details for
Quantity
  • Availability
  • Add to Cart