ENA388700536 Enamine

1-amino-3-[2-(methylsulfanyl)ethyl]thiourea

  • MDL number MFCD19618227
  • Cas Number : 77780-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124580
Availability details for
Quantity
  • Availability
  • Add to Cart