ENA388700539 Enamine

2-{6-bromo-3H-imidazo[4,5-b]pyridin-2-yl}pyridine

  • MDL number MFCD09750275
  • Cas Number : 537029-58-6
  • Molecular Weight : 275.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122883
Availability details for
Quantity
  • Availability
  • Add to Cart