ENA388700540 Enamine

4-bromo-N-{[4-(trifluoromethyl)phenyl]methyl}aniline

  • MDL number MFCD12443795
  • Cas Number : 1041538-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124587

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-124587
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart