ENA388700550 Enamine

7,8-difluoro-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD12785665
  • Cas Number : 1249025-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-124627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124627
Availability details for
Quantity
  • Availability
  • Add to Cart