ENA388700552 Enamine

2-(3-chlorophenoxy)-3-methylbutanoic acid

  • MDL number MFCD11127004
  • Cas Number : 1019372-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124630

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124630
Availability details for
Quantity
  • Availability
  • Add to Cart