ENA388700553 Enamine

4-(3,4-dimethoxyphenyl)-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD23144231
  • Cas Number : 1427379-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124632
Availability details for
Quantity
  • Availability
  • Add to Cart