ENA388700559 Enamine

1-(pyridin-2-yl)-1H-1,2,4-triazol-3-amine

  • MDL number MFCD14594798
  • Cas Number : 1307174-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124639
Availability details for
Quantity
  • Availability
  • Add to Cart