ENA388700560 Enamine

2-[2-(hydroxymethyl)phenyl]ethan-1-ol

  • MDL number MFCD19300992
  • Cas Number : 6346-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124640
Availability details for
Quantity
  • Availability
  • Add to Cart