ENA388700641 Enamine

2-(6-aminopyridin-3-yl)-2-methylpropanoic acid hydrochloride

  • MDL number MFCD22741798
  • Cas Number : 246219-78-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124651

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124651
Availability details for
Quantity
  • Availability
  • Add to Cart