ENA388700647 Enamine

5-(aminomethyl)-3-(2-methylpropyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD23144238
  • Cas Number : 1427380-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124673
Availability details for
Quantity
  • Availability
  • Add to Cart