ENA388700656 Enamine

3-[(2R)-pyrrolidin-2-yl]propanoic acid hydrochloride

  • MDL number MFCD23144248
  • Cas Number : 188415-35-2
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-3-(pyrrolidin-2-yl)propanoic acid hydrochloride
Inchi Code 1S/C7H13NO2.ClH/c9-7(10)4-3-6-2-1-5-8-6;/h6,8H,1-5H2,(H,9,10);1H/t6-;/m1./s1
Inchi Key FDACNBQRVTXPOH-FYZOBXCZSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124722
Availability details for
Quantity
  • Availability
  • Add to Cart