ENA388700661 Enamine

1-(1-benzofuran-2-yl)-3-(4-methylpiperazin-1-yl)propan-1-one dihydrochloride

  • MDL number MFCD23144255
  • Cas Number : 1427380-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124741

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124741
Availability details for
Quantity
  • Availability
  • Add to Cart