ENA388700666 Enamine

4-ethyl-N-phenyl-4H-1,2,4-triazol-3-amine

  • MDL number MFCD23144263
  • Cas Number : 1427379-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124788
Availability details for
Quantity
  • Availability
  • Add to Cart