ENA388700678 Enamine

spiro[2.5]octan-1-amine hydrochloride

  • MDL number MFCD23144279
  • Cas Number : 17202-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124870
Availability details for
Quantity
  • Availability
  • Add to Cart