ENA388700688 Enamine

3-[2-(pyrrolidin-1-yl)ethyl]aniline dihydrochloride

  • MDL number MFCD23144290
  • Cas Number : 1427380-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124915

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124915
Availability details for
Quantity
  • Availability
  • Add to Cart