ENA388700690 Enamine

1,2,3-trimethylpiperazine

  • MDL number MFCD23144292
  • Cas Number : 1427378-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124962
Availability details for
Quantity
  • Availability
  • Add to Cart