ENA388700692 Enamine

2-amino-2-(4-chloro-3-fluorophenyl)ethan-1-ol hydrochloride

  • MDL number MFCD23144294
  • Cas Number : 1427380-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124984

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124984
Availability details for
Quantity
  • Availability
  • Add to Cart