ENA388700696 Enamine

benzyl(dodecan-2-yl)amine

  • MDL number MFCD23144302
  • Cas Number : 1427378-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125068

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125068
Availability details for
Quantity
  • Availability
  • Add to Cart