ENA388700698 Enamine

2-methyl-3-(morpholin-4-yl)aniline dihydrochloride

  • MDL number MFCD23144304
  • Cas Number : 1427380-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125080

Safety Information

msds https://www.enaminestore.com/catalog/EN300-125080
Availability details for
Quantity
  • Availability
  • Add to Cart