ENA388700704 Enamine

N-(3-amino-4-fluorophenyl)-2-(4-ethylpiperazin-1-yl)acetamide dihydrochloride

  • MDL number MFCD23144310
  • Cas Number : 1427379-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-125156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-125156
Availability details for
Quantity
  • Availability
  • Add to Cart