ENA388700717 Enamine

5-chlorohexanoyl chloride

  • MDL number MFCD20623759
  • Cas Number : 99585-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124789
Availability details for
Quantity
  • Availability
  • Add to Cart