ENA388700718 Enamine

2,2,2-trifluoro-1-(1-phenyl-1H-pyrazol-4-yl)ethan-1-ol

  • MDL number MFCD16215894
  • Cas Number : 1275913-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 77...79
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124790

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124790
Availability details for
Quantity
  • Availability
  • Add to Cart