ENA388700721 Enamine

N-cyclopentylaminosulfonamide

  • MDL number MFCD16878609
  • Cas Number : 866490-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124665

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124665
Availability details for
Quantity
  • Availability
  • Add to Cart