ENA388700727 Enamine

N-[(4-methoxyphenyl)methyl]methanesulfonamide

  • MDL number MFCD01212931
  • Cas Number : 42060-31-1
  • Molecular Weight : 215.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124801
Availability details for
Quantity
  • Availability
  • Add to Cart