ENA388700728 Enamine

2-fluoro-6-iodo-4-methylaniline

  • MDL number MFCD15527690
  • Cas Number : 217314-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 32...34
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124805

Safety Information

msds https://www.enaminestore.com/catalog/EN300-124805
Availability details for
Quantity
  • Availability
  • Add to Cart